Goat anti-Guinea Pig IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555 - FAQs
Goat anti-Guinea Pig IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555 - FAQs
View additional product information for Goat anti-Guinea Pig IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555 - FAQs (A21435)
1 product FAQs found
My Goat anti-Guinea Pig IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor 555 (Cat. No. A21435) was shipped at ambient temperature even though the recommended storage is at 2-4 degrees C. Will this affect the stability or performance?
Goat anti-Guinea Pig IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor 555 (Cat. No. A21435) is routinely shipped at room temperature and this does not affect stability or performance of the product.