Amplex™ Red Catalase Assay Kit
Invitrogen™

Amplex™ Red Catalase Assay Kit

For use with absorbance and fluorescence intensity techniques
Have Questions?
Catalog NumberQuantity
A22180400 Assays
Catalog number A22180
Price (USD)
526.00
Each
Quantity:
400 Assays
Price (USD)
526.00
Each

The Amplex Red Catalase Assay Kit provides a sensitive and simple fluorometric method for detecting as little as 50 mU/mL of catalase activity in a purified system in a 100μL assay volume.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Detection MethodAbsorbance
Dye TypeAmplex Red
Emission Wavelength585 nm
Excitation Wavelength571 nm
Quantity400 Assays
Shipping ConditionRoom Temperature
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase) Substrate, Catalase Substrate
Target EnzymeCatalase
For Use With (Application)Enzyme and Metabolic Activity
Product LineAmplex
Product TypeCatalase Assay
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?

The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can Amplex Red Assays be performed using cell lysates?

This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.

The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (12)

Citations & References
Abstract
RIP1 potentiates BPDE-induced transformation in human bronchial epithelial cells through catalase-mediated suppression of excessive reactive oxygen species.
Authors:Wang Q, Chen W, Xu X, Li B, He W, Padilla MT, Jang JH, Nyunoya T, Amin S, Wang X, Lin Y,
Journal:Carcinogenesis
PubMed ID:23633517
'Cell survival signaling is important for the malignant phenotypes of cancer cells. Although the role of receptor-interacting protein 1 (RIP1) in cell survival signaling is well documented, whether RIP1 is directly involved in cancer development has never been studied. In this report, we found that RIP1 expression is substantially increased ... More
Physiological characterization of Haemophilus influenzae Rd deficient in its glutathione-dependent peroxidase PGdx.
Authors:Pauwels F, Vergauwen B, Van Beeumen JJ
Journal:J Biol Chem
PubMed ID:14701867
'The chimeric peroxidase PGdx of Haemophilus influenzae Rd belongs to a recently identified family of thiol peroxidases capable of reducing hydrogen peroxide as well as alkylhydroperoxides by means of glutathione redox cycling. In the present study, we constructed a H. influenzae Rd strain, deficient in its PGdx encoding gene (open ... More
Receptor-interacting Protein 1 Increases Chemoresistance by Maintaining Inhibitor of Apoptosis Protein Levels and Reducing Reactive Oxygen Species through a microRNA-146a-mediated Catalase Pathway.
Authors:Wang Q, Chen W, Bai L, Chen W, Padilla MT, Lin AS, Shi S, Wang X, Lin Y,
Journal:
PubMed ID:24425875
'Although receptor-interacting protein 1 (RIP1) is well known as a key mediator in cell survival and death signaling, whether RIP1 directly contributes to chemotherapy response in cancer has not been determined. In this report, we found that, in human lung cancer cells, knockdown of RIP1 substantially increased cytotoxicity induced by ... More
Roles of catalase and hydrogen peroxide in green tea polyphenol-induced chemopreventive effects.
Authors:Yamamoto T, Lewis J, Wataha J, Dickinson D, Singh B, Bollag WB, Ueta E, Osaki T, Athar M, Schuster G, Hsu S
Journal:J Pharmacol Exp Ther
PubMed ID:14569057
'The green tea polyphenol (-)-epigallocatechin-3-gallate (EGCG) possesses promising anticancer potential. Although in vivo studies unveiled the metabolic routes and pharmacokinetics of EGCG and showed no adverse effects, in vitro studies at high concentrations demonstrated oxidative stress. EGCG causes differential oxidative environments in tumor versus normal epithelial cells, but the roles ... More
N-acetylcysteine reverses cardiac myocyte dysfunction in a rodent model of behavioral stress.
Authors:Chen F, Hadfield JM, Berzingi C, Hollander JM, Miller DB, Nichols CE, Finkel MS,
Journal:
PubMed ID:23722706
Compelling clinical reports reveal that behavioral stress alone is sufficient to cause reversible myocardial dysfunction in selected individuals. We developed a rodent stress cardiomyopathy model by a combination of prenatal and postnatal behavioral stresses (Stress). We previously reported a decrease in percent fractional shortening by echo, both systolic and diastolic ... More