RapidFinder™ Rabbit ID Kit
Thermo Scientific™
RapidFinder™ Rabbit ID Kit
The Thermo Scientific™ RapidFinder™ Rabbit ID Kit enables the simple and reliable detection of rabbit (Oryctolagus cuniculus) DNA in food and feed samples.
Have Questions?
카탈로그 번호수량
A2440848 reactions
카탈로그 번호 A24408
제품 가격(KRW)
-
수량:
48 reactions
The RapidFinder™ Rabbit ID Kit enables the simple and reliable detection of rabbit (Oryctolagus cuniculus) DNA in food and feed samples.
The RapidFinder™ Rabbit ID Kit enables the simple and reliable detection of Rabbit DNA in food and feed samples. The method works by detection of rabbit (Oryctolagus cuniculus) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Rabbit ID Kit detects and provides sensitivity down to 0.01% of Rabbit DNA. The included positive control in the kit, which comprises 0.1% Rabbit DNA, can be used as a reference to set the threshold of detection of the assay.
  • Simple- Real-time PCR based assay, no electrophoresis needed
  • Flexible- Rabbit DNA detected in any food or feed sample, both raw and processed
  • Reliable- Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast- From sample to result in the same day
The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Real-Time PCR Systems.
Sample Preparation
DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.
Part of a complete portfolio of Solutions for Animal Species Detection
The RapidFinder™ Rabbit ID Kit is just one of our many available kits for the sensitive detection of various meat species in food and feed samples. Other RapidFinder™ meat species identification kits available include those for the detection of pork, beef, horse, goat, water buffalo, fallow deer, duck, sheep, ruminant, chicken, poultry, and turkey.
The RapidFinder™ Species Identification Kits are made available through a partnership with Health in Code, S.L.
식품 및 환경 시료 측정 전용.
사양
설명Rabbit ID Kit
용도(장비)QuantStudio 5™ and 7500 Fast
제품라인RapidFinder
제품 유형RapidFinder™
수량48 reactions
샘플 종류Food and Environmental Samples
민감도0.01% of Rabbit (Oryctolagus cuniculus) DNA
유통 기한2 years
Oryctolagus cuniculus
검출 방법Primer-probe
용도(애플리케이션)Food and Beverage
PCR 방법qPCR
반응 속도Standard
구성 및 보관
Contains 1 tube each of: Rabbit Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)