RapidFinder™ Specific Tuna ID Kit
Thermo Scientific™
RapidFinder™ Specific Tuna ID Kit
The Thermo Scientific™ RapidFinder™ Specific Tuna ID Kit enables the simple and reliable detection of 4 types of tuna DNA in food and feed samples.
Have Questions?
Número de catálogoCantidad
A2441148 reactions
Número de catálogo A24411
Precio (USD)
-
Cantidad:
48 reactions
The Thermo Scientific ™ RapidFinder™ Specific Tuna ID Kit enables real-time PCR detection of specific tuna DNA that is present in food and feed samples. The kit detects blends containing ≥1% of the following tuna species through the specific amplification of a specific region present in the mitochondrial genome: Thunnus alalunga (Longfin tuna), Thunnus albacares (Yellowfin tuna), Thunnus obesus (Bigeye tuna), and Katsuwonus pelamis (Skipjack tuna).
The RapidFinder™ Specific Tuna ID Kit enables the simple and reliable detection of tuna DNA in food and feed samples. The method works by detection of specific tuna (Thunnus alalunga, Thunnus albacares, Thunnus obesus, and Katsuwonus pelamis) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Specific Tuna ID Kit detects blends containing ≥1% of the following tuna species through the specific amplification of a specific region present in the mitochondrial genome: Thunnus alalunga, Thunnus albacares, Thunnus obesus, and Katsuwonus pelamis DNA.
  • Simple-real-time PCR based assay, no electrophoresis needed
  • Flexible-specific tuna DNA detected in any food or feed sample, both raw and processed
  • Reliable-Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast-from sample to result in the same day
The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Food Safety Real-Time PCR systems.
Sample Preparation
DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.
Part of a Complete Portfolio of Solutions for Fish Species Detection
The RapidFinder™ Pollack ID Kit is just one of our many available kits for the sensitive detection of various fish species in food and feed samples. Other RapidFinder™ Fish species identification kits available include those for the detection of Basa, Gadus, Haddock, Pollock, Pollack, and Tuna.
Únicamente para las pruebas de muestras alimentarias y ambientales.
Especificaciones
DescripciónSpecific Tuna ID Kit
Para utilizar con (equipo)QuantStudio 5™ and 7500 Fast
Línea de productosRapidFinder
Cantidad48 reactions
Tipo de muestraFood and Environmental Samples
Sensibilidad1% of Specific Tuna: Thunnus alalunga, Thunnus albacares, Thunnus obesus, and Katsuwonus pelamis DNA
Duración de almacenamiento2 years
EspecieThunnus alalunga, Thunnus albacares, Thunnus obesus, and Katsuwonus pelamis
Método de detecciónPrimer-probe
Para utilizar con (aplicación)Food and Beverage
Método de PCRqPCR
Velocidad de reacciónStandard
Unit SizeEach
Contenido y almacenamiento
Contains 1 tube each of: Master Mix Tuna 1 (store at -20°C) Master Mix Tuna 2 (store at -20°C) Positive Control Tuna 1 (1%) (store at -20°C)  Positive Control Tuna 2 (1%) (store at -20°C)  and 2 tubes of General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use)