Product Image
Applied Biosystems™
HID Real-Time PCR Analysis Software v1.2 Upgrade
HID Real-Time PCR Analysis Software is a tool for forensic laboratories to optimize their quantification workflow and enhance their downstream decision making process.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A246145 Licenses
A246651 licencia
Número de catálogo A24614
Precio (MXN)
-
Cantidad:
5 Licenses
HID Real-Time PCR Analysis Software is a tool for forensic laboratories to optimize their quantification workflow and enhance their downstream decision making process.
The software has the following workflow features:
  • customizable sample templates
  • advanced data analysis capabilities
  • normalization tool
  • STR setup tool
HID Real-Time PCR Analysis Software v1.2 is designed for the Applied Biosystems™ 7500 Real-Time PCR System. It offers a set of features beyond instrument controls. It streamlines the experimental setup, supports customizable workflow templates and enhances the data analysis and review process. The software is validated for use on both the Windows™ XP and Windows™ 7 platforms, and it is specialized for all available Quantifiler™ kits, including the Quantifiler™ Trio and the Quantifiler™ HP DNA Quantification kits.
This product includes a HID Real-Time PCR Analysis Software v1.2 Upgrade DVD and 1 user license to upgrade from v1.1 of the software. Documentation is not included.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Para utilizar con (aplicación)PCR en tiempo real (PCRq)
Para utilizar con (equipo)Sistema 7500
Rendimiento de PCR enriquecido por GCLow
Cantidad5 Licenses
Tipo de productoHID Real Time PCR Analysis Software
Unit SizeEach
Preguntas frecuentes
How does the quantification solution provided by the Quantifiler Trio and HP DNA Quantification Kits and HID Real-Time PCR Software easily integrate with robotics for an automated workflow?
Robotic integration is enabled by the automation-friendly bundling of the Quantifiler Trio and HP DNA Quantification kits combined with the HID Real-Time PCR Analysis Software, which has been designed to facilitate automation with commonly used robotic platforms. In addition, our Human Identification Professional Services (HPS) team has extensive expertise in developing customized robotic scripts and conducting on-site verification and validation testing to help you achieve optimal performance with a wide range of platforms. The HPS team has already developed scripts for the Quantifiler Trio and HP DNA Quantification kits on several of the most common robotic platforms, and would be happy to consult with your lab on how to efficiently implement these solutions into your workflow. Please contact your local human identification (HID) account manager to learn more.
Is HID Real-Time PCR Software v1.4 compatible with data files generated by the previous software versions?
Yes, the HID Real-Time PCR Analysis Software v1.4 can read and reanalyze data files generated by the previous versions of HID Real-Time PCR Analysis Software.
Is the HID Real-Time PCR Analysis Software v1.4 compatible with all previously released Quantifiler kits?
Yes, v1.4 does work with previously released Quantifiler kits.
How does the HID Real-Time PCR Software v1.4 differ from the previous version, v1.3?
HID Real-Time PCR Software v1.4 includes the same feature set as version 1.3 and runs on Windows 10 as well. The difference is that HID Real-Time PCR software v1.4 is designed to be compatible with the latest QuantStudio 5 firmware version 1.5.1.
How is the HID Real-Time PCR Software different from the basic SDS software?
The HID Real-Time PCR Software has been fully validated for human identification. The key workflow features include configurable Quantifiler templates as well as instrument calibration, data quality assessment, and integrated sample normalization/STR reaction setup tools for simplified, faster sample processing from quantification to STR analysis. It can automatically calculate the degradation index, male:female mixture ratio, and dilution values, and includes a number of customizable data quality flags for rapid data review. The HID Real-Time PCR Software uses different analysis algorithms than the SDS v1.2.3 software. In our internal comparison testing, the HID Real-Time PCR Software generated CT values with statistically significant differences compared to the SDS v1.2.3 software.