The Attune™ Cytometric Software versions 5.1.1 and 5.2 must use the English version OS of Windows 10 with language and regional settings set to English.
Product Image
Thermo Scientific™
Attune™ NxT Software, single license
Attune™ NxT Software is used to operate and maintain the Attune™ NxT acoustic focusing cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A24855
also known as A-24855
10 Licenses
A24856
also known as A-24856
5 Licenses
Catalog number A24855
also known as A-24855
Price (USD)
-
Quantity:
10 Licenses
Attune™ NxT Software is used to operate and maintain the Attune™ NxT acoustic focusing cytometer. It enables powerful user-defined experimental analysis via an intuitive, easy-to-use interface and provides advanced tools for both acquisition and analysis that are simple enough for users at all experience levels.
  • Simplified use—streamlined, intuitive interface
  • Speed—faster real-time analysis of large data sets
  • Easy instrument operation—automated maintenance and instrument performance monitoring
Simplified use
Attune™ NxT software employs a menu system that presents the features you need when you need them without a lot of scrolling and clicking. The customization tab allows you to pre-define settings for plots, gates, experiments, reports, and overlays. To simplify experimental set up, the preview screen shows all plot combinations instantaneously allowing you to easily add them to the experiment workspace. Compensation is automated or user-defined and can be set up using a compensation guide.
Speed increases productivity
Attune™ NxT Software has been designed to maximize the efficiency in performing data analysis. It uses a Windows™ 7 64-bit system to provide fast refresh rates for large data sets of up to 20 million events created by multi-laser, multi-parameter instrumentation. The software allows you to immediately visualize changes to your data plots in real-time as you make adjustments. This allows you to spend less time analyzing data and more time on your research.
Easy instrument operation
Flow cytometers often require the user to spend time ensuring the quality of the system through maintenance tasks and proper QC, since changes in lasers, fluidics, and optics can affect the quality of the data generated. Attune™ NxT Software takes many of these manual tasks and fully automates them to help reduce time spent on instrument maintenance. The software provides an automated quality control instrument tracking system, important for collecting and analyzing accurate data requiring minimal user interaction.
Software features:
  • Instrument controls
  • Automated maintenance and performance testing
  • Data collection
  • Visualization tools
  • Dot plot, density plot, histogram
  • Histogram marker, quad marker
  • Customized logic gates
  • Data plot preview for quick plot set-up
  • Data display—log, linear, log transforms
  • Statistics—mean, median, mode, %CV, rCV, SD, % total, % gated
  • Experiment set-up
  • Automated compensation
  • Auto layout and freeform workspace layouts
  • Analysis tools
  • Multi-file selection tools
  • File management
  • Import/export of files and experiments
  • Files stored as FCS3.1 format
  • User management & security
  • User and administrator accounts—basic/admin
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Attune™ NxT Acoustic Focusing Cytometer
Product LineAttune
Product TypeFlow Cytometer Software
Quantity10 Licenses
Shipping ConditionRoom Temperature
Software Type10 individual user licenses
Unit Size10 licenses
Contents & Storage
10 Attune™ NxT Software dongles, 10 copies Attune™ NxT Software