Invitrogen™

Attune™ NxT Flow Cytometer, blue, refurbished

Catalog number: A24864R
Invitrogen™

Attune™ NxT Flow Cytometer, blue, refurbished

Catalog number: A24864R
Catalog Number
A24864R
Unit Size
1 cytometer
Price (USD)
Catalog NumberUnit SizePrice (USD)
A24864R1 cytometer
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
This is a refurbished Attune NxT Flow Cytometer, which is an easy-to-use instrument designed to detect cell size and granularity, as well as fluorescence expression of labeled antibodies, proteins, and functional reagents. The innovative design of the instrument results in precise data and acquisition speeds up to 10X faster than other flow cytometers. The design also eliminates lengthy sample preparation procedures, reduces the time to analyze a file, allows small or limited samples to be analyzed, and minimizes variability in measurements, leading to better resolution of populations. It is ideal for immunophenotyping and signaling studies, cell cycle analysis, detection of rare events, stem cell analysis, cancer and apoptosis studies, and microbiological assays.

With the Attune NxT Flow Cytometer you get:
• A flexible platform that provides highly sensitive and accurate measurement of cell sizeand granularity, as well as fluorescence detection
• State of the art optics for long-term reliability
• Acoustic focusing for high-precision data
• An intuitive software user interface that reduces time to proficiency
• Improved workflow to reduce sample preparation time before analysis
• Faster data analysis that leads to reduced acquisition times

Flexible design that accommodates changing needs, budget, or laboratory space
The instrument's modular design accommodates the most common fluorophores and fluorescent proteins used in flow cytometry enabling alignment with previous experimental results and accommodating future needs through simple upgrades. The small footprint of the instrument requires minimal bench space, even allowing use in a cell culture hood.

Acoustic focusing improves efficiencies compared to traditional flow cytometers
Traditional flow cytometers rely on a large volume of fluid to focus cells prior to analysis, resulting in extensive sample preparation prior to analysis and introducing the possibility of error due to artifacts created during the sample preparation process. With the Attune NxT Flow Cytometer, blood samples can be run without being manipulated using the No-Wash No-Lyse blood cell assay. The instrument also offers flexibility in the sample concentration that can be analyzed.

Acoustic focusing enables:
• Elimination of concentration steps
• Effective background fluorescence reduction by a simple dilution step, eliminating the need to wash fluorescence samples before analysis
• No-lyse/no-wash assays
• Analysis of samples that have low cell numbers, including CSF, stem cells, mouse bone marrow, mouse blood, and thin needle aspirates
• Acquisition of a high volume of samples in minimal time
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Tube(s), 384-well plate, 96-well plate
Laser Features
Blue Excitation Laser (488 nm)
Product Line
Attofluor™
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice

Contents & Storage

• Attune Acoustic Focusing Cytometer, refurbished
• Minitower Windows™ 7 64-bit computer with 23-inch flat panel monitor
• Power cord with adapters
• USB cable
• Attune NxT software
• Quick reference card
• 1 vial of Attune Performance Tracking Beads
• 1 Bottle of Attune Wash Solution
• 1 Bottle of Attune 1 x Shutdown Solution
• Attune Focusing Fluid

Figures