TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging
Invitrogen™
TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging
The Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits are the only kits offering superior imaging for liveRead more
Have Questions?
Catalog NumberQuantity
A2487950 tests
Catalog number A24879
Price (INR)
71,648.00
Each
Quantity:
50 tests
Price (INR)
71,648.00
Each
The Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits are the only kits offering superior imaging for live human PSCs in one box. This TRA-1-60 Alexa Fluor™ 555 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the PSC marker TRA-1-60. For fixed cell staining of PSC markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit.
With the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging, you can:
• Confirm the expression of a key pluripotent stem cell marker using an optimally dye-conjugated antibody
• Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM imaging medium
• Stain live cell samples with confidence using reagents that are tested to be free of common contaminants
Confirm Expression of a Key PSC Marker
Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM imaging medium.
Preserve Cell Health and Maximize Signal
Staining cells in FluoroBrite™ DMEM imaging medium maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.
Stain Live-cell Samples with Confidence
Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2 μm sterile-filtered and tested to be free of common contaminants.
What You Get
Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 uL staining volume and includes:
• 200 μL of Alexa Fluor™ 555 Mouse Anti-Human TRA-1-60 (50X concentration)
• 500 mL FluoroBrite™ DMEM
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeConjugate Kit
Quantity50 tests
ConjugateAlexa Fluor 555
Unit SizeEach
Contents & Storage
  • 200 μL of Alexa Fluor™ 555 Mouse Anti-Human TRA-1-60 (0.25 mg/mL) (store at 2°C to 8°C)
  • 500 mL FluoroBrite™ DMEM
  • Store at 2°C to 8°C, protect from light