The QuantStudio™ 3D Digital PCR Tilt Base for ProFlex™ Thermal Cycler sits beneath the ProFlex™ Thermal Cycler and ensures consistent performance during PCR amplification of the QuantStudio™ 3D Digital PCR chips. Simply affix the two risers and place the tilt base beneath the feet of the thermal cycler, load your prepared 3D Digital PCR chips, and you are ready for PCR amplification.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
ProFlex™ PCR System, QuantStudio™ 3D PCR System
Product Type
3D Digital PCR Tilt Base
For Use With (Application)
Real Time PCR (qPCR)
Contents & Storage
Includes: 1 tilt base, 2 tilt base risers, and 12 riser cable ties