Consistone™ Yeast Extract UF
Gibco™

Consistone™ Yeast Extract UF

The first functionally defined protein hydrolysate, delivering peptone-level productivity with CD-like predictability, helping ensure consistent performance, faster scale-up, and greater confidence in upstream bioprocesses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A253050110 kgPail
A2530502500 gBottle
Catalog number A2530501
Price (MXN)
-
Quantity:
10 kg
Format:
Pail

Gibco Consistone Yeast Extract, ultra-filtered (UF), is a functionally defined protein hydrolysate designed for CHO applications. It combines the productivity benefits of traditional peptones with the predictability of chemically defined media, helping teams achieve consistent performance and faster process optimization. With controlled trace-metal levels, Consistone extract supports smoother DoE and scale-up, helping reduce risk, improve reproducibility, and give early-stage development programs the confidence to advance with speed and reliability.

Consistone Yeast Extract UF features include:

  • Highly consistent performance with 10–20% lower lot-to-lot variability versus industry benchmarks for more predictable runs and fewer repeats
  • Enhanced quality control, including bioburden ≤300 CFU/g and every-lot endotoxin testing ≤500 EU/g
  • Proven productivity gains, delivering >20% higher titer vs. TC Yeastolate UF in CHO cells
  • Optimized trace-metal ranges for consistent growth, glycosylation, enzyme cofactor control, and stable titer
  • Animal origin–free, supporting global regulatory acceptance and helping reduce compliance risk
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian, Insect, Microbial
Culture TypeMammalian, Insect, Microbial
Endotoxin Level≤500 EU/g
For Use With (Application)Preparing and supplementing bacterial, insect, and mammalian cell culture media
FormatPail
Manufacturing QualityUF
Quantity10 kg
ClassificationAnimal Origin-free, Serum-free
FormPowder
Product TypeCell Culture Supplement
SterilityNon-sterile
With AdditivesNone
Without AdditivesNo Animal Derived Components
Unit SizeEach