Introducing Cells-to-CT™ Express 1-Step! Discover Cells-to-CT™ Express 1-Step, our next-generation workflow designed for faster, simpler gene expression analysis.
Cells-to-CT™ 1-Step TaqMan™ Kit
Invitrogen™

Cells-to-CT™ 1-Step TaqMan™ Kit

Like other members of the Cells-to-CT™ product family, the Cells-to-CT™ 1-Step TaqMan™ Kit makes it possible to perform expression analysisRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A2560520 Reactions
A25603100 Reactions
A25602400 Reactions
Catalog number A25605
Price (USD)
433.65
Online exclusive
477.00
Save 43.35
Each
No. of Reactions:
20 Reactions
Price (USD)
433.65
Online exclusive
477.00
Save 43.35
Each

Like other members of the Cells-to-CT™ product family, the Cells-to-CT™ 1-Step TaqMan™ Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification.

This new addition to the family includes a TaqMan™ 1-Step RT-PCR master mix. The single-tube format of the RT-PCR step streamlines the 'sample to answer' workflow more than ever. Combining the simple Cells-to- CT™ lysis technology with the fast cycling capability of the 1-step RT-PCR master mix allows quicker time to results with limited sample handling.

The Cells-to-CT™ 1-Step TaqMan™ Kit is:

  • Complete—optimized workflow includes cell lysis reagents with gDNA removal and TaqMan™ 1-Step RT-PCR Master Mix
  • Faster than ever—7-minute sample prep, including DNase treatment, combined with a 28-minute real-time RT-PCR step allows for ∼35 minute sample-to-answer workflow
  • Simple—10 steps total (sample prep and real-time RT-PCR combined)
  • Robust—perform gene expression analysis on 10–100,000 cells per sample; results equivalent to those from purified RNA
  • Convenient—contains sufficient reagents to perform 5 real-time RT-PCRs for each cell sample you prepare; this allows you to study multiple targets or perform replicate RT-PCRs

Note: TaqMan™ primer/probe sets are sold separately.

Simple 7-minute Sample Preparation, Then Ready for Real-time RT-PCR

Whether you are using plates or tubes, the Cells-to-CT™ 1-Step TaqMan™ Kit uses a simple 7-minute sample preparation procedure. The lysis technology is designed for 10–100,000 cultured cells/sample. Cells are washed in PBS and lysed in solution for 5 minutes at room temperature with a simultaneous DNase treatment. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution. The lysate is now ready to analyze on your real-time PCR instrument. Simply add it to a real-time RT-PCR master mix containing the TaqMan™ primer/probe sets of interest. Alternatively, the lysate can be stored at –20°C. Because samples can be processed directly in culture wells (96 or 384 wells), sample handling and the potential for sample loss or transfer error are minimized, facilitating rapid, high-throughput processing. Unlike old-fashioned, multi-step RNA isolation protocols, no heating, washing, or centrifugation steps are required. The kit greatly simplifies a laborious 30–60 minute process and reduces it to 7 minutes.

A Complete, Validated Solution

All components of the Cells-to-CT™ 1-Step TaqMan™ Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate sample preparation and real-time 1-step RT-PCR master mixes. For added quality assurance, the Cells-to-CT™ 1-Step TaqMan™ Kit has been validated with TaqMan™ Gene Expression assays and shows performance equivalent to that obtained with purified RNA.

Accessory Product

The TaqMan™ Cells-to-CT™ Control Kit was designed for use with TaqMan™ Cells-to-CT™ products. This kit contains an endogenous control (ACTB) to normalize for sample loading variability and an artificial XenoRNA™ control and corresponding TaqMan™ Gene Expression assay to monitor the effects of PCR inhibition.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio™, ViiA™ 7 System, StepOnePlus™ System, StepOne™ System, Real-Time PCR Systems or Thermal Cyclers, 7500 Fast Dx System, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
FormatTube
No. of Reactions20 Reactions
Product LineCells-to-CT, TaqMan
Product Type1-Step qPCR kit
Quantity20 preps
Reagent TypeCell Lysis
Sample TypeMammalian Cells
Starting Material10-100,000 Cells
Volume2.2 mL
Detection MethodTaqMan
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR Method1-step RT-qPCR
Reaction Speed7 min.
Unit SizeEach
Contents & Storage
Lysis Solution, 2.2 mL, store at 2°C to 8°C, DNase I, 22 μL, store at -5 to -30°C, Stop Solution, 200 μL, store at -5 to -30°C, TaqMan™ 1-step qRT-PCR Mix, 250 μL, store at -5 to -30°C,

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.