Countess™ Reusable Cell Counting Chamber Slide and Holder, no stain, 1 glass slide - FAQs
Countess™ Reusable Cell Counting Chamber Slide and Holder, no stain, 1 glass slide - FAQs
View additional product information for Countess™ Cell Counting Chamber Slides - FAQs (A40008469, C10283, C10315, C10314, A25750, C10228, C10313, C10312)
3 product FAQs found
Do you offer reusable slides and the appropriate holders for Countess II FL and Countess 3/3 FL instruments?
The Countess Reusable Slide (Cat. No. A25750) is compatible with the Countess II FL and the Countess 3/3 FL instruments. However, they require a reusable slide holder specific to the instrument. The Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208) and the Countess II FL Reusable Slide Holder (Cat No. AMEP4746) are shipped with the instrument and can be purchased separately.
The slide holder can be held inside the back of the instrument, next to the light cube removal tool.
Which cover slips can I use with the Countess Reusable Slide (Cat. No. A25750)?
The Hausser Scientific Cover Glasses/Slips (Cat. No. 02617581) from Fisher Scientific are compatible with the with the Countess Reusable Slide (Cat. No. A25750).
Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center
What Countess Automated Cell Counter instruments are compatible with the Countess Reusable Slide (Cat. No. A25750)?
The Countess Reusable Slide (Cat. No. A25750) is compatible with Countess 2 FL, Countess 3, and Countess 3 FL Automated Cell Counter instruments.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center .