Cassette Clamp, left
Cassette Clamp, left
Cassette clamps are replacement parts for the Mini Gel Tank. They lock and seal the gel cassettes within the GelLeia mais
Have Questions?
Número do catálogoQuantity
A259461 unit
Número do catálogo A25946
Preço (BRL)
-
Quantity:
1 unit
Cassette clamps are replacement parts for the Mini Gel Tank. They lock and seal the gel cassettes within the Gel Runner Tank. The orientation is determined when facing the front of the gel tank.
About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Quantity1 unit
Shipping ConditionRoom Temperature
For Use With (Equipment)Universal
Gel CompatibilityNovex™ Mini Gels
Gel SizeMini
TypeCassette Clamp
Unit SizeEach
Frequently asked questions (FAQs)
Can I purchase replacement parts for the Mini Gel Tank?
Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the components of the Mini Gel Tank?
The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.