PSC Cryopreservation Kit
Gibco™

PSC Cryopreservation Kit

The PSC Cryopreservation Kit contains xeno-free PSC Cryopreservation Medium, which is a ready-to-use solution for the cryopreservation of early passage자세히 알아보기
Have Questions?
카탈로그 번호수량
A264460150 mL
카탈로그 번호 A2644601
제품 가격(KRW)
457,000
온라인 행사
Ends: 30-Sep-2026
507,000
할인액 50,000 (10%)
Each
수량:
50 mL
제품 가격(KRW)
457,000
온라인 행사
Ends: 30-Sep-2026
507,000
할인액 50,000 (10%)
Each
The PSC Cryopreservation Kit contains xeno-free PSC Cryopreservation Medium, which is a ready-to-use solution for the cryopreservation of early passage pluripotent stem cells (PSCs), and RevitaCell™ Supplement (100X), a chemically defined recovery supplement for use in the post-thaw culture media. When used in combination, these reagents help minimize loss of cell viability, maximize post-thaw recovery, and minimize unwanted differentiation of pluripotent stem cells. This kit can also be used to cryopreserve and recover peripheral blood mononuclear cells (PBMCs) to improve post-thaw cell viability and recovery.

• Efficient recovery—routinely achieves 50% more viable cells than traditional methods of PSC cryopreservation
• Consistent—chemically defined media maintains normal morphology, pluripotency, and karyotype of pluripotent stem cells over 10 passages
• Reduced variability—optimized reagents minimize the causes of genetic instability of pluripotent stem cells

Reliable
Xeno-free, chemically defined formulations help ensure the proliferation capacity and long-term culture of PSCs and PBMCs in a stable state, thus helping to increase their usefulness in research and clinical research applications.

Easy to use
The PSC Cryopreservation Medium is provided as a ready-to-use, complete solution containing 10% DMSO for efficient cell addition. Separate additions of enhancing reagents are not required, helping save valuable preparation time.

Versatile
The kit is compatible with cells cultured in a variety of media systems including Essential 8™ medium, StemPro™ hESC SFM, and other commercially available formulations. Also, the kit is compatible with clump and single cell passaged PSCs.

cGMP manufacturing and quality system
The PSC Cryopreservation Kit is manufactured at a cGMP-compliant facility. The facility is also registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
사양
제조 품질cGMP for medical devices, 21 CFR Part 820 and ISO 13485
제품라인Gibco
제품 유형PSC Cryopreservation Kit
수량50 mL
배송 조건Dry Ice
분류Xeno-free for Cryomedium, Animal Origin-free for RevitaCell
Culture TypeStem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
Serum LevelSerum-free
첨가제 포함DMSO (10%)
Unit SizeEach
구성 및 보관
• 50 mL Cryopreservation Media
• 5 mL RevitaCell Supplement (100X)

Store at -5 to -20°C and protect from light.

자주 묻는 질문(FAQ)

Can RevitaCell Supplement be used in addition to a traditional ROCK Inhibitor?

No. Combination of RevitaCell Supplement with a traditional ROCK inhibitor will result in deleterious effects on PSCs.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

The RevitaCell Supplement in the PSC Cryopreservation Kit has a strong smell. Is this normal?

Yes, it is normal that the RevitaCell Supplement has a strong smell.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between the PSC Cryopreservation Kit and the CTS PSC Cyropreservation Kit?

The CTS PSC Cryopreservation Kit (Cat. No. A4239301) provides the same efficient recovery, consistency, and reduced variability as the PSC Cryopreservation Kit (Cat. No. A2644601). However, these two kits have different intended use statements, i.e., “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals” and “For Research Use Only”, respectively. Furthermore, the CTS PSC Cryopreservation Kit has full USP sterility testing info and a Drug Master file (DMF), whereas the sterility testing of the PSC Cryopreservation is via a modified USP method, and it does not have a DMF.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I cryopreserve the iPSCs?

You can cryopreserve iPSCs just as you would cryopreserve any pluripotent stem cells. Growth medium with 10% DMSO is recommended for freezing, or our PSC cryopreservation kit (Cat. No. A2644601) can be used

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can the Cryopreservation Medium from PSC Cryopreservation Kit be aliquoted and frozen again?

We have not tested this. However, the medium is stable when stored at 4 degrees C for up to 6 months. There are no components that R&D would be concerned about during a freeze thaw; however, this was not formally tested.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.