Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
CTS™ Essential 8™ Medium
Gibco™

CTS™ Essential 8™ Medium

CTS Essential 8 Medium is a fully-defined, feeder-free medium designed for the growth and expansion of hPSCs for clinical applications and formulated with components not directly derived from animals.
Have Questions?
Catalog NumberQuantity
A2656101500 mL
Catalog number A2656101
Price (USD)
480.00
Each
Quantity:
500 mL
Price (USD)
480.00
Each

Gibco CTS Essential 8 Medium is a fully-defined, feeder-free medium designed for the growth and expansion of hPSCs for clinical applications and formulated with components not directly derived from animals. The formulation of CTS Essential 8 Medium is based on Essential 8™ Medium, which was originally developed by Guokai Chen et al. in the laboratory of James Thomson (published as 'E8'). CTS Essential 8 Medium provides similar performance to Essential 8 Medium, supporting the transition from the bench to the clinic.

CTS Essential 8 Medium:

  • Reduces risks with a fully defined formulation
  • Simplifies the transition from bench to clinic with similar performance and the same 8-component formulation as RUO Essential 8 Medium, but with components not directly derived from animals
  • Facilitates customer regulatory filings with cGMP manufacturing, traceability of raw materials, and available documentation, including certificates of origin, drug master files, and regulatory support

Reduces risks from animal-origin material

CTS Essential 8 Medium contains no animal- or human-derived components at the primary component level. It is a fully defined medium for pluripotent stem cell culture.

Simplifies transition from bench to clinic

Like RUO Essential 8 Medium, CTS Essential 8 Medium contains only the eight essential components needed for stem cell culture, but with components not directly derived from animals. Pluripotent stem cells (PSCs) cultured in CTS Essential 8 Medium have been shown to maintain PSC marker expression after long-term culture and to generate cells of the three germ lineages.

Combined with cGMP manufacturing, this simple, fully defined formulation improves consistency compared to other media that contain over 20 highly variable ingredients. The close similarity between Essential 8 Medium and CTS Essential 8 Medium supports the transition from one medium to the other.

Facilitates regulatory filings

Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available for CTS Essential 8 Medium.

CTS Essential 8 Medium is commercialized in partnership with Cellular Dynamics International.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeEmbryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBottle
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product TypeCell Culture Medium
Quantity500 mL
Shelf Life12 months from date of manufacture
Shipping ConditionCTS Essential 8 Medium: Ambient, CTS Essential 8 Supplement: Dry Ice
ClassificationXeno-free
FormLiquid
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
• 500 mL CTS Essential 8 Medium; store at 2–8°C and protect from light
• 10 mL CTS Essential 8 Supplement; store at –5 to –20°C and protect from light

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

At what points in induced pluripotent stem (iPS) cell culture should CTS RevitaCell Supplement (100X) (Cat. No. A4238401) be used?

CTS RevitaCell Supplement (100X) should be used in combination with cell culture medium as a post-thaw recovery solution to improve cell viability. It can also be used in combination with CTS Essential 8 Medium (Cat. No. A2656101) when performing single cell passaging procedures.

Find additional tips, troubleshooting help, and resources within our Stem Cell Culture Support Center