Product Image
Applied Biosystems™
3500 Dx Data Collection Software 3, Upgrade Kit
3500 Dx Series Data Collection Software 3 provides the ideal features and functionality for use in both clinical and researchMás información
Have Questions?
Número de catálogoCantidad
A278551 Kit
Número de catálogo A27855
Precio (MXN)
-
Cantidad:
1 Kit
3500 Dx Series Data Collection Software 3 provides the ideal features and functionality for use in both clinical and research environments. Powered by the Windows™ 7 operating system, the 3500 Dx Series Data Collection Software 3 provides:
• Dual-mode user interface, the Diagnostic Mode and the Research Use Only Mode
• Enables both fragment analysis and Sanger sequencing in the secure Diagnostic Mode
• Enables import of validated sequencing and fragment analysis assay protocols to run in Diagnostic Mode
• Comprehensive dashboard that delivers regular updates of instrument status and remaining days of onboard consumables
• SAE (Security, Audit, and Electronic) signature capability to assist with compliance to governing regulations
This product includes a computer system, monitor, 3500 Dx Series Data Collection Software 3 upgrade USB flash drive, one licensing kit with single license code, and two user document USB flash drives.
For an upgrade, a full install and operational and performance qualification package by a certified engineer is required. These services are ordered and provided separately.
Especificaciones
Para utilizar con (aplicación)Secuenciación
Para utilizar con (equipo)Analizador genético 3500 Dx
Tipo de productoData Collection Software
Cantidad1 Kit
Condiciones de envíoTemperatura ambiente
FormatoKit
Unit SizeEach
Contenido y almacenamiento
• Dell Workstation and monitor
• 3500 Dx Series Data Collection Software 3 USB
• 1 licensing kit with single license code
• 2 user document USBs