LiquidBiopsy™ Blood Collection Kit

Catalog number: A28171

Ion Torrent™  Related applications: Digital PCR | Ion Torrent™ Next-Generation Sequencing | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The LiquidBiopsy™ Blood Collection Kit contains a fixative and an easy-to-use protocol to stabilize blood samples for up to 96 hours enabling ambient temperature storage and shipping of blood samples prepared for automated rare cell recovery on the LiquidBiopsy™ Instrument.

Easy-to-use protocol
Stabilizing blood samples prior to processing for cfDNA or CTC research requires only a few minutes of hands-on time; the protocol is complete in a little more than an hour.

1. Prepare a working fixative solution for a 10 mL blood sample by gently mixing two provided solutions.
2. Allow the fixative solution to stand at room temperature for 15 minutes to activate. Once activated, the solution must be used within 24 hours.
3. Pour the 10 mL blood sample from the EDTA tube into the fixative solution. Secure tightly and invert three times to mix.
4. Allow the sample to sit undisturbed for 1 hour. The specimen is stable for up to 96 hours. No cooling required.

For best results, the blood sample should be fixed within 2 hours of collection and maintained at ambient temperature until fixation.

The LiquidBiopsy™ Blood Collection Kit is part of the LiquidBiopsy™ Platform for automated rare cell isolation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: DNA
For Use With (Equipment): Ion PGM™ System
Product Line: LiquidBiopsy™
Product Size: 1 reaction
Sequencing Type: Genome & DNA Sequencing
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products