LiquidBiopsy™ Reagents and Consumables Kit, Part 1

Catalog number: A28186

Ion Torrent™  Related applications: Digital PCR | Ion Torrent™ Next-Generation Sequencing | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The LiquidBiopsy™ Reagents and Consumables Kit is a two-part kit (each part sold separately) that includes sufficient buffers, reagents, vials, elution tubes, and flow cells for the processing of up to eight blood samples using the LiquidBiopsy™ Instrument for automated rare cell recovery.

Part 1 (A28186) contains mostly plastic/glass consumables (vials, filter tips, non-filter tips, elution tubes, flow cell covers, flow cells, reagent reservoirs, sealing gaskets) plus LiquidBiopsy™ Bottom Buffer. These items are stored at room temperature. Reagent reservoirs and LiquidBiopsy™ Bottom Buffer expire three months after manufacture.

Part 2 (A28187) contains reagents (beads, antibodies, wash and reagent buffers). They are stored at variable temperatures (see product manual) and expire three months after manufacture.

Analyze samples using EpCAM or an antibody cocktail
The full kit includes the LiquidBiopsy™ flow cell that captures circulating tumor cells (CTCs) using a proprietary immunocapture protocol and one EpCAM antibody for automated CTC capture using positive selection. Researchers may choose to develop an antibody cocktail for blood samples containing tumor types with low levels of EpCAM expression.

The LiquidBiopsy™ Blood Collection Kit is part of the LiquidBiopsy™ Platform for automated rare cell isolation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: DNA
For Use With (Equipment): Ion PGM™ System
Product Line: LiquidBiopsy™
Product Size: 8 reactions
Sequencing Type: Genome & DNA Sequencing
Shipping Condition: Room Temperature

Contents & storage

Vials, filter tips, non-filter tips, elution tubes, flow cell covers, flow cells, reagent reservoirs, sealing gaskets, and LiquidBiopsy™ Bottom Buffer.

Store contents at room temperature. Reagent reservoirs and LiquidBiopsy™ Bottom Buffer expire after three months.

Documents

Manuals & protocols

Recommended products