ProteinSEQ™ Protein A Core Kit
Applied Biosystems™

ProteinSEQ™ Protein A Core Kit

The ProteinSEQ™ Protein A Core Kit is a partial kit that enables users to customize their ProteinSEQ protein measurements forRead more
Have Questions?
Catalog NumberQuantity
A28406200 Reactions
Catalog number A28406
Price (USD)
-
Quantity:
200 Reactions
The ProteinSEQ™ Protein A Core Kit is a partial kit that enables users to customize their ProteinSEQ protein measurements for leeched Protein A, or any other ligand used in their purification columns. It contains the buffers and reagents, but not the antibody-loaded capture beads, needed to enable PCR-based quantitation of Protein A or any other purification ligand. The kit contains sufficient reagents for 200 reactions. For a kit that includes Protein A capture beads and standards, for use in the quantitation of Protein A, please see the ProteinSEQ™ Protein A Quantitation Kit (Cat. No. 4469343).

• Quantitate leeched protein contaminants with ultra-high sensitivity
• Accelerate process development with more valid test results per run
• Reduce assay hands-on time with less sample dilution and preparation
• Achieve unprecedented productivity with near-5-log assay dynamic range

ProteinSEQ Protein A quantitation
The ProteinSEQ assay platform uses qPCR technology to measure process contaminants with extraordinary sensitivity and unparalleled dynamic range. Protein A or other purification ligands leeching from purification columns can be accurately measured with outstanding spike recovery and dilutional linearity.

Streamlined assay worflow
The ProteinSEQ system is the only instrument platform-based Protein A and purification ligand quantitation solution to offer single-digit picogram sensitivity and nearly 5 logs of dynamic range. Laboratory workflow is greatly accelerated due to the reduction in the number of sample dilutions and replicates in each run.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayProtein Quantitation Assay
For Use With (Application)Proximity Ligation Assay
FormatKit
Quantity200 Reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Product LineProteinSEQ
TypeProtein A Core Kit
Unit Size200 reactions
Contents & Storage
Store at 4–8°C:
• ProteinSEQ elution buffer
• Fast master mix (2X)
• Wash buffer
• ProteinSEQ diluent

Store at -5 to -30°C:
• ProteinSEQ ligation & assay mix (20X)
• ProteinSEQ ligase
• Protein A sample prep reagent (2X)