Product Image
Applied Biosystems™
3500 Dx Series Data Collection Software 3
3500 Dx Series Data Collection Software 3 offers features and functionality for use in both clinical and research environments. PoweredMás información
Have Questions?
Número de catálogoCantidad
A288691 kit
Número de catálogo A28869
Precio (MXN)
-
Cantidad:
1 kit
3500 Dx Series Data Collection Software 3 offers features and functionality for use in both clinical and research environments. Powered by the Windows™ 7 operating system, the 3500 Dx Series Data Collection Software 3 provides:
• Dual-mode user interface, the Diagnostic Mode and the Research Use Only Mode
• Enables both fragment analysis and Sanger sequencing in the secure Diagnostic Mode
• Enables import of validated sequencing and fragment analysis assay protocols to run in Diagnostic Mode
• Comprehensive dashboard that delivers regular updates of instrument status and remaining days of onboard consumables
• SAE (Security, Audit, and Electronic) signature capability to assist with compliance with governing regulations
This product includes a 3500 Dx Series Data Collection Software 3 USB flash drive, one licensing kit with single license code, and two user document USB flash drives. This product does not include a computer, installation reagents, or the qualification service. For a regular upgrade, please refer to the upgrade kit and qualification service.
Especificaciones
Para utilizar con (equipo)Analizador genético 3500 Dx
Tipo de producto3500 Dx Series Data Collection Software 3
Cantidad1 kit
Condiciones de envíoTemperatura ambiente
Software TypeUSB, License Required, CE-IVD Regulated
Unit SizeEach
Contenido y almacenamiento
• 3500 Dx Series Data Collection Software 3 USB
• 1 licensing kit with single license code
• 2 user document USBs