PSC Cardiomyocyte Maintenance Medium
Gibco™

PSC Cardiomyocyte Maintenance Medium

Gibco™ Cardiomyocyte Maintenance Medium is a serum-free and xeno-free medium that is capable of maintaining cardiomyocytes that have been differentiated자세히 알아보기
Have Questions?
카탈로그 번호수량
A2920801500 mL
카탈로그 번호 A2920801
제품 가격(KRW)
269,000
온라인 행사
Ends: 30-Jun-2026
298,000
할인액 29,000 (10%)
Each
수량:
500 mL
제품 가격(KRW)
269,000
온라인 행사
Ends: 30-Jun-2026
298,000
할인액 29,000 (10%)
Each
Gibco™ Cardiomyocyte Maintenance Medium is a serum-free and xeno-free medium that is capable of maintaining cardiomyocytes that have been differentiated using the PSC Cardiomyocyte Differentiation Kit (Cat. No. A2921201). This medium is included in the kit and sold separately for researchers wishing to maintain differentiated cardiomyocytes in culture for extended periods of time.

• No thawing or mixing required—simply warm and add to differentiated cardiomyocyte culture
• Long-term culture—maintain differentiated cardiomyocytes in culture for >15 days
• Stable phenotype—maintain phenotype throughout long-term culture experiments
For Research Use Only. Not for use in diagnostic procedures.
사양
세포 유형Cardiomyocytes
제품 유형Maintenance Medium
수량500 mL
배송 조건Room Temperature
Culture TypeMammalian Cell Culture
Serum LevelSerum-free
Unit SizeEach

자주 묻는 질문(FAQ)

What substrate should I use for cardiomyocyte differentiation?

We recommend using Gibco Geltrex Flex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A4000046801, A4000046802, or A4000046803) or Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A400457 or A400458) for xeno-free applications.