VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay
The VetMAX™ Xeno™ Internal Positive Control (IPC) - LIZ™ Assay is a primer-probe mix that detects the Xeno internal positiveRead more
Have Questions?
Change view
Catalog Number
No. of Reactions
A29766
100
A29768
500
2 Options
Catalog number A29766
Price (USD)
187.65
Online exclusive
208.00
Save 20.35
Each
No. of Reactions:
100
Price (USD)
187.65
Online exclusive
208.00
Save 20.35
Each
The VetMAX™ Xeno™ Internal Positive Control (IPC) - LIZ™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results. The Xeno IPC LIZ assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.
Features of the VetMAX Xeno IPC - LIZ Assay include: • Provides confidence that qPCR test results are accurate and actionable • Easily integrates into any workflow • Greatly reduces the likelihood of false negatives
The VetMAX Xeno IPC - LIZ Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno™ Internal Positive Control RNA or VetMAX™ Xeno™ Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.
Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay (Cat. No. A29765) is also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Detection and Identification of Animal Disease
IncludesInternal Positive Control Assay
Label or DyeLIZ
No. of Reactions100
Product LineVetMAX, Xeno
Product TypeInternal Positive Control
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesNot Species-Specific
FormatKit
Unit SizeEach
Contents & Storage
1 x 110 μL Xeno LIZ Primer Probe Mix. Store at -5°C to -30°C.