CleanSweep™ PCR Purification Reagent
Applied Biosystems™
CleanSweep™ PCR Purification Reagent
The CleanSweep™ PCR Purification Reagent is used for enzymatic cleanup of an amplified PCR product. The reagent hydrolyzes excess primersMás información
Have Questions?
Número de catálogoCantidad
A298985000 Reactions
Número de catálogo A29898
Precio (USD)
-
Cantidad:
5000 Reactions
The CleanSweep™ PCR Purification Reagent is used for enzymatic cleanup of an amplified PCR product. The reagent hydrolyzes excess primers and nucleotides in one single step. CleanSweep reagent-purified samples are ready for use in downstream applications such as DNA sequencing or single nucleotide polymorphism (SNP) analysis.
Request a free sample >
Remove unincorporated primers and nucleotides enzymatically
Prepare DNA sequencing samples with one pipetting step
Conserve precious PCR products with 100% recovery
Eliminate column cleanup entirely
The CleanSweep one-step solution provides a simple way to enzymatically purify a PCR reaction. The reaction is complete with one pipetting step followed by two incubations. The first incubation hydrolyzes excess primer and dephosphorylates nucleotides. The second high temperature incubation inactivates the enzymes.
Enzymatic cleanup is the preferred method for minute amounts of sample such as PCR product. The PCR amplicons are completely conserved, as there are no transfer steps to other tubes, wells, or columns required. At the same time, cross-contamination is limited.
No further processing is needed. After enzymatic cleanup with CleanSweep reagent, primers and dNTPs will no longer interfere with DNA sequencing or SNP analysis.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Compatibilidad de alto rendimientoHigh-throughput Compatible, Manual Protocols, Multiplexing, Automated Protocols
N.º de reacciones5000 reactions
Línea de productosCleanSweep
Tipo de productoPCR Purification Reagent
Diana de purificaciónADN de reacciones enzimáticas (p. ej PCR)
Cantidad5000 Reactions
Tipo de muestraAny DNA Sample Source
Condiciones de envíoDry Ice
Para utilizar con (aplicación)Standard PCR, Cloning
Unit Size5000 reactions
Contenido y almacenamiento
Almacenar en congelador (de -5 a -30 °C).