Product Image
Ion Torrent™
TherapyEdge™ Box Server, academic/government
The TherapyEdge™ Box server includes pre-installed and pre-configured software that makes it easy for a research facility of any sizeMás información
Have Questions?
Número de catálogoCantidad
A29989A
también denominado A29989A 
1 servidor
Número de catálogo A29989A
también denominado A29989A 
Precio (MXN)
-
Cantidad:
1 servidor
The TherapyEdge™ Box server includes pre-installed and pre-configured software that makes it easy for a research facility of any size or at any level of sophistication to instantly begin viral typing research studies. It has been developed with a specific focus for HIV monitoring, but because of its extensible architecture it has been extended for monitoring of other viruses including hepatitis B and hepatitis C.
The TherapyEdge Box integrates easily into any network architecture and thus provides a single point of access to numerous knowledge bases. Because of its web-based interface and architecture, one TherapyEdge box can serve a single facility or a large geography with numerous remote facilities such a regional research site. This makes it a cost-effective solution. The TherapyEdge Box is built using open source technologies, making it very extensible and robust without incurring significant ownership costs.
The TherapyEdge system offers the following features:
• Linux-based operating system
• Web browser interface to access local server
• Data stored in MySQL database
• Connectivity and real-time maintenance
• DSL, 3G, dial-up
• DVD-ROM
• IT Administration Interface
• Research sample identification with fingerprint
• Several types of user levels (show/edit/delete, restriction on pools of data)
• Encrypted hard drive
• Secured SSL connections
• Firewall
• SSH remote connections (authentication key)
• RAID
• Backup on DVD, through Internet, on external hard drives
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Tipo de productoBox Server
Cantidad1 servidor
Condiciones de envíoTemperatura ambiente
Línea de productosTEBox
Unit SizeEach
Contenido y almacenamiento
1 TEBox Server (academic/government)