Now available:Upgraded Alexa Fluor Antibody Labeling kits with user-friendly, advanced purification columns that ensure rapid dye removal and high yield of labeled proteins in just 30 minutes, with minimal hands-on time.
Alexa Fluor™ 594 Microscale Protein Labeling Kit
Invitrogen™

Alexa Fluor™ 594 Microscale Protein Labeling Kit

Microscale Protein Labeling Kits provide a convenient means for attaching a fluorescent label to a small amount of antibody orRead more
Have Questions?
Catalog NumberQuantity
A300081 kit
Catalog number A30008
Price (USD)
798.00
Each
Quantity:
1 kit
Price (USD)
798.00
Each
Microscale Protein Labeling Kits provide a convenient means for attaching a fluorescent label to a small amount of antibody or protein (20–100 μg). The kits are available in four Alexa Fluor™ colors (or biotin) and supply everything needed for three labeling and separation reactions.

Important Features of Microscale Protein Labeling Kits:
• Labeled proteins typically ready to use typically in 2 hours (∼30 minutes hands-on time)
• Optimized for 20–100 μg of protein with molecular weights between 12 and 150 kDa
• Purified using convenient spin filters with yields between 60 and 90%
• Stabilizing proteins must be removed from the sample before labeling

Stable Reaction Chemistry and Superior Alexa Fluor™ Dyes
In the Microscale Protein Labeling Kits, the reactive dye contains a succinimidyl (NHS) ester moiety that reacts with primary amines of proteins to form stable dye-protein conjugates. Compared to traditional dyes, Alexa Fluor™ dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. And generally when using Alexa Fluor™ dyes, higher degrees of labeling can be achieved without intramolecular quenching. For details see Alexa Fluor™ Dyes Spanning the Visible and Infrared Spectrum—Section 1.3.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.

We’ll Make a Custom Antibody Conjugate for You
If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
Detection MethodFluorescence
Excitation/Emission590/617 nm
Label TypeAlexa Fluor
Labeling MethodConjugation-based
Labeling Scale20–100 μg
Product LineAlexa Fluor
Product TypeLabeling Kit
Quantity1 kit
Reactive MoietySuccinimidyl (NHS) Ester
Shipping ConditionRoom Temperature
Labeling TargetProteins
Label or DyeAlexa Fluor 594
Unit SizeEach
Contents & Storage
Store in refrigerator 2°C to 8°C and protect from light.

Frequently asked questions (FAQs)

Can I use 50 μg of protein with Fluorescent Protein Labeling Kits?

No. We recommend using 1 mg of protein with Fluorescent Protein Labeling Kits. For smaller protein sample sizes, we recommend using Microscale Protein Labeling kits which are optimized for 20-100 µg of protein.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What formulation of antibody should I use for conjugation for small animal in vivo imaging?

To allow for good reaction kinetics, antibodies should be in PBS buffer at a concentration of 0.5-3.0 mg/ml. The antibody must be free of preservatives (azide etc.), amine containing buffers and carrier proteins such as BSA.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is degree of labeling (DOL)?

Degree of labeling (DOL) describes the number of fluorophores per antibody. For in vivo labeling experiments, the DOL is restricted to a narrow range because it has significant consequences for the biodistribution and clearance of the probe. For example, for in vivo imaging, we have determined that the DOL range for the far-red Alexa Fluor dyes is 1.5 to 3 molecules per antibody for optimal optical in vivo imaging.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (3)

Citations & References
Abstract
Focal targeting by human ß-defensin 2 disrupts localized virulence factor assembly sites in Enterococcus faecalis.
Authors:Kandaswamy K, Liew TH, Wang CY, Huston-Warren E, Meyer-Hoffert U, Hultenby K, Schröder JM, Caparon MG, Normark S, Henriques-Normark B, Hultgren SJ, Kline KA,
Journal:
PubMed ID:24191013
'Virulence factor secretion and assembly occurs at spatially restricted foci in some Gram-positive bacteria. Given the essentiality of the general secretion pathway in bacteria and the contribution of virulence factors to disease progression, the foci that coordinate these processes are attractive antimicrobial targets. In this study, we show in Enterococcus ... More
Serum amyloid A3 binds MD-2 to activate p38 and NF-?B pathways in a MyD88-dependent manner.
Authors:Deguchi A, Tomita T, Omori T, Komatsu A, Ohto U, Takahashi S, Tanimura N, Akashi-Takamura S, Miyake K, Maru Y,
Journal:
PubMed ID:23858030
Serum amyloid A (SAA) 3 is a major component of the acute phase of inflammation. We previously reported that SAA3 served as an endogenous peptide ligand for TLR4 to facilitate lung metastasis. Because these experiments were performed with SAA3 recombinant proteins purified from Escherichia coli or mammalian cells, we could ... More
LytA, major autolysin of Streptococcus pneumoniae, requires access to nascent peptidoglycan.
Authors:Mellroth P, Daniels R, Eberhardt A, Rönnlund D, Blom H, Widengren J, Normark S, Henriques-Normark B,
Journal:J Biol Chem
PubMed ID:22334685
The pneumococcal autolysin LytA is a virulence factor involved in autolysis as well as in fratricidal- and penicillin-induced lysis. In this study, we used biochemical and molecular biological approaches to elucidate which factors control the cytoplasmic translocation and lytic activation of LytA. We show that LytA is mainly localized intracellularly, ... More