Now available:Upgraded Alexa Fluor Antibody Labeling kits with user-friendly, advanced purification columns that ensure rapid dye removal and high yield of labeled proteins in just 30 minutes, with minimal hands-on time.
Alexa Fluor™ 647 Microscale Protein Labeling Kit
Invitrogen™

Alexa Fluor™ 647 Microscale Protein Labeling Kit

Microscale Protein Labeling Kits provide a convenient means for attaching a fluorescent label to a small amount of antibody orRead more
Have Questions?
Catalog NumberQuantity
A300091 Kit
Catalog number A30009
Price (USD)
798.00
Each
Quantity:
1 Kit
Request bulk or custom format
Price (USD)
798.00
Each

Microscale Protein Labeling Kits provide a convenient means for attaching a fluorescent label to a small amount of antibody or protein (20–100 μg). The kits are available in four Alexa Fluor™ colors (or biotin) and supply everything needed for three labeling and separation reactions.

Important Features of Microscale Protein Labeling Kits:

  • Labeled proteins typically ready to use typically in 2 hours (∼30 minutes hands-on time)
  • Optimized for 20–100 μg of protein with molecular weights between 12 and 150 kDa
  • Purified using convenient spin filters with yields between 60 and 90%
  • Stabilizing proteins must be removed from the sample before labeling

Stable Reaction Chemistry and Superior Alexa Fluor™ Dyes

In the Microscale Protein Labeling Kits, the reactive dye contains a succinimidyl (NHS) ester moiety that reacts with primary amines of proteins to form stable dye-protein conjugates. Compared to traditional dyes, Alexa Fluor™ dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. And generally when using Alexa Fluor™ dyes, higher degrees of labeling can be achieved without intramolecular quenching. For details see Alexa Fluor™ Dyes Spanning the Visible and Infrared Spectrum—Section 1.3.

Learn More About Protein and Antibody Labeling

We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.

We'll Make a Custom Antibody Conjugate for You

If you can't find what you're looking for in our stocked list, we'll prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorFar-Red
Detection MethodFluorescence
Excitation/Emission650/665 nm
Label TypeAlexa Fluor
Labeling MethodConjugation-based
Labeling Scale20–100 μg
Product LineAlexa Fluor
Product TypeLabeling Kit
Quantity1 Kit
Reactive MoietySuccinimidyl (NHS) Ester
Shipping ConditionRoom Temperature
Labeling TargetProteins
Label or DyeAlexa Fluor 647
Unit SizeEach
Contents & Storage
Store in refrigerator 2°C to 8°C and protect from light.

Frequently asked questions (FAQs)

Can I use 50 μg of protein with Fluorescent Protein Labeling Kits?

No. We recommend using 1 mg of protein with Fluorescent Protein Labeling Kits. For smaller protein sample sizes, we recommend using Microscale Protein Labeling kits which are optimized for 20-100 µg of protein.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What formulation of antibody should I use for conjugation for small animal in vivo imaging?

To allow for good reaction kinetics, antibodies should be in PBS buffer at a concentration of 0.5-3.0 mg/ml. The antibody must be free of preservatives (azide etc.), amine containing buffers and carrier proteins such as BSA.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is degree of labeling (DOL)?

Degree of labeling (DOL) describes the number of fluorophores per antibody. For in vivo labeling experiments, the DOL is restricted to a narrow range because it has significant consequences for the biodistribution and clearance of the probe. For example, for in vivo imaging, we have determined that the DOL range for the far-red Alexa Fluor dyes is 1.5 to 3 molecules per antibody for optimal optical in vivo imaging.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (4)

Citations & References
Abstract
Ubiquitination screen using protein microarrays for comprehensive identification of Rsp5 substrates in yeast.
Authors:Gupta R,Kus B,Fladd C,Wasmuth J,Tonikian R,Sidhu S,Krogan NJ,Parkinson J,Rotin D
Journal:Molecular systems biology
PubMed ID:17551511
Ubiquitin-protein ligases (E3s) are responsible for target recognition and regulate stability, localization or function of their substrates. However, the substrates of most E3 enzymes remain unknown. Here, we describe the development of a novel proteomic in vitro ubiquitination screen using a protein microarray platform that can be utilized for the ... More
Mouse marginal zone B cells harbor specificities similar to human broadly neutralizing HIV antibodies.
Authors:Pujanauski LM, Janoff EN, McCarter MD, Pelanda R, Torres RM,
Journal:Proc Natl Acad Sci U S A
PubMed ID:23288906
'A series of potent, broadly neutralizing HIV antibodies have been isolated from B cells of HIV-infected individuals. VRC01 represents a subset of these antibodies that mediate neutralization with a restricted set of IGHV genes. The memory B cells expressing these antibodies were isolated years after infection; thus, the B-cell subpopulation ... More
Microstructure, local viscoelasticity and cell culture suitability of 3D hybrid HA/collagen scaffolds.
Authors:Roether J, Bertels S, Oelschlaeger C, Bastmeyer M, Willenbacher N
Journal:PLoS One
PubMed ID:30566463
'As mechanical properties of cell culture substrates matter, methods for mechanical characterization of scaffolds on a relevant length scale are required. We used multiple particle tracking microrheology to close the gap between elasticity determined from bulk measurements and elastic properties sensed by cells. Structure and elasticity of macroporous, three-dimensional cryogel ... More
Microtubule-Actomyosin Mechanical Cooperation during Contact Guidance Sensing.
Authors:Tabdanov ED, Puram V, Zhovmer A, Provenzano PP
Journal:Cell Rep
PubMed ID:30304674
Cancer cell migration through and away from tumors is driven in part by migration along aligned extracellular matrix, a process known as contact guidance (CG). To concurrently study the influence of architectural and mechanical regulators of CG sensing, we developed a set of CG platforms. Using flat and nanotextured substrates ... More