MicroAmp™ Optical 8-Tube Strip with Attached Optical Caps, 0.2 mL - FAQs
MicroAmp™ Optical 8-Tube Strip with Attached Optical Caps, 0.2 mL - FAQs
View additional product information for MicroAmp™ Optical 8-Tube Strip with Attached Optical Caps, 0.2 mL - FAQs (A30588)
6 product FAQs found
Are the Tubes, 0.2 mL, flat cap (Cat. No. AB0620) also available in a strip format, i.e., as multiple tubes attached to each other?
Yes. Please see the following catalog numbers: AB1182, AB1114, and A30588.
Do you offer an alternative for MicroAmp Optical 8-Tube Strip with Attached Optical Caps, 0.2 mL (Cat. No. A30588)?
We recommend using the following qPCR and PCR Plastics Selection Tool for finding an alternative: https://www.thermofisher.com/de/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html
What are the benefits of using MicroAmp Optical 8-Tube Strip with Attached Optical Caps?
These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Is the MicroAmp Optical 8-Tube Strip with Attached Optical Caps DNase free?
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Which instrument(s) is the MicroAmp Optical 8-Tube Strip with Attached Optical Caps compatible with?
The MicroAmp Optical 8-Tube Strip with Attached Optical Caps is compatible with the following instruments:
- ProFlex 3 x 32-well PCR System
- ProFlex 96-well PCR System
- Veriti 96-well Thermal Cycler
- SimpliAmp Thermal Cycler
- MiniAmp Thermal Cycler
- MiniAmp Plus Thermal Cycler
- 2720 Thermal Cycler
- GeneAmp PCR System 9700, 96-well
- 7000 PCR System
- 7300 Real-Time PCR System
- 7500 Real-Time PCR system
- QuantStudio Real-Time PCR Systems
- ViiA7 Real-Time PCR System, 96-well
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Do I need to use a tray with the MicroAmp Optical 8-Tube Strip with Attached Optical Caps?
MicroAmp 96-Well Tray for VeriFlex Blocks (Cat. No. 4379983) is needed to load 1 or 2 MicroAmp Optical 8-Tube Strips with Attached Optical Caps on the ProFlex, SimpliAmp, MiniAmp, MiniAmp Plus, and Veriti Thermal Cycler 96-well blocks, and QuantStudio 3 and QuantStudio 5 blocks.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.