Does the JetFlex Genomic DNA Purification Kit work with frozen blood?
Yes. However, the yield from frozen blood is significantly lower than the yield from fresh blood.
Search
View additional product information for JetFlex™ Genomic DNA Purification Kit - FAQs (A30700)
1 product FAQs found