Thermo Scientific™

POROS™ MabCapture™ A RoboColumn

Catalog number: A30726
Thermo Scientific™

POROS™ MabCapture™ A RoboColumn

Catalog number: A30726
Catalog Number
A30726
Unit Size
600 µL
Price (USD)
950.00
Your Price:
Availability
***
Quantity
Catalog Number
Unit Size
200 µL
Price (USD)
318.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A30726600 µL
950.00
Your Price:
***
A30725200 µL
318.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS™ MabCapture™ A affinity resin, which is suitable for monoclonal antibody large-scale downstream bioprocessing.

POROS MabCapture A is a 50-micron affinity resin that combines improvements made in bead and chemistry technology. These advances provide a monoclonal antibody purification resin that not only demonstrates the highest dynamic binding performance at low flow rates, but also one that can maintain superior binding capacity at high linear velocities where other chromatography resins products cannot operate.

Features of the resin:
• Obtain the highest dynamic binding capacity for monoclonal antibodies vs. conventional agarose or silica based Protein A medias
• Maintain high dynamic binding capacity at flow rates over 700 cm/hr
• Design processes with greater flexibility (shorter bed heights, faster flow rates or both) due to the improvements made to the antibody binding efficiency
• Clean and sanitize with standard sodium hydroxide agents utilized for other chromatography products

RoboColumn is a registered trademark of ATOLL GmbH.

Specifications

Column Type
Affinity
Product Line
MABCAPTURE™, POROS™
Purification Target
Protein
Shipping Condition
Wet Ice
Stationary Phase
Protein A
Product Type
Column
Quantity
600 µL
Format
Chromatography Cartridge

Contents & Storage

Store at 2–8°C.

Figures

Frequently asked questions (FAQs)

Citations & References