Bacto™ Yeast Extract 2
Gibco™
Bacto™ Yeast Extract 2
Bacto Yeast Extract 2 is a concentrate of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A30801-01500 g
A30801-0210 kg
A30801-0350 kg
Catalog number A30801-01
Price (MXN)
-
Quantity:
500 g
Bacto Yeast Extract 2 is a concentrate of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. Derived from primary grown baker's yeast, this animal-origin free product is suitable for use in microbial fermentation. Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation.
Bacto Yeast Extract 2 is one of the most complete and versatile fermentation supplements available.
Related yeast extract products
  • Gibco™ Yeast Extract – developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
  • Difco Yeast Extract UF – ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g.
  • Difco Yeast Extract Low-Dusting (LD) – designed to limit the amount of dust generated compared to other spray-dried yeast extracts
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Culture TypeMammalian, Microbial, and Insect
FormatBottle
Quantity500 g
Shelf Life1095 days
ClassificationAnimal Origin-free
FormPowder
Product TypeYeast Supplement
SterilityNon-sterile
Unit Size500 g