POP-7™ Polymer for Protein Quality Analysis
POP-7™ Polymer for Protein Quality Analysis
Applied Biosystems™

POP-7™ Polymer for Protein Quality Analysis

This is a pouch of POP-7 polymer for protein quality analysis on 3500/3500xL genetic analyzers. The POP-7 separation matrix isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A30936384 Samples
A31122960 Samples
Catalog number A30936
Price (USD)/ Each
-
Request A Quote
Quantity:
384 Samples
This is a pouch of POP-7 polymer for protein quality analysis on 3500/3500xL genetic analyzers. The POP-7 separation matrix is validated for protein quality analysis and is offered in easy-to-use pouch packages and 960 sample (maximum) or 384 sample (maximum) size formats.

• Easy to use—pre-formulated and in a single-use pouch, which not only saves time but ensures reliability and consistency
• Includes radio frequency identification (RFID) label—easily track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
• Robust formulation—formulated for biomolecular analysis applications
• Replenishable—allows capillaries to be used multiple times

Reproducible results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help ensure reproducibility. POP polymers are specifically formulated to separate biomolecules of known size range at a desired resolution and run time.

For partially-used pouches that need to be stored, a cap that plugs into the pouch opening is recommended (Cat. No. 4412619).
For research use only
Specifications
DescriptionPOP-7™ Polymer For 3500xL Genetic Analyzer
For Use With (Application)Glycan Analysis, Protein Quality Analysis
For Use With (Equipment)3500xL Genetic Analyzer for Protein Quality Analysis
No. of Samples384
Product LinePOP-7™
Product TypePolymer
Quantity384 Samples
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
Store at 2–8°C.