POP-7™ Polymer for Protein Quality Analysis, 960 samples - FAQs
POP-7™ Polymer for Protein Quality Analysis, 960 samples - FAQs
View additional product information for POP-7™ Polymer for Protein Quality Analysis - FAQs (A31122)
1 product FAQs found
What is the difference between the compositions of POP-7 in POP-7 Polymer for Protein Quality Analysis (Cat. Nos. A30936, A31122) and POP-7 Polymer, for 3500/SeqStudio Flex (Cat. Nos. 4393708, A26703, 4393714)?
There is no difference between the compositions of POP-7 in POP-7 Polymer for Protein Quality Analysis (Cat. Nos. A30936, A31122) and POP-7 Polymer, for 3500/SeqStudio Flex (Cat. Nos. 4393708, A26703, 4393714) as they come from the same manufacturing bulk/formulation.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.