IonCode™ Barcode Adapters 1-384 Kit (for Agrigenomics) - FAQs
IonCode™ Barcode Adapters 1-384 Kit (for Agrigenomics) - FAQs
View additional product information for IonCode™ Barcode Adapters 1-384 Kit (for Agrigenomics) - FAQs (A31173)
1 product FAQs found
Where can I find the sequences for the IonCode barcodes?
The sequences for the barcodes can be found in the Barcodes section of your Torrent Browser. You can also download a .csv file containing the full list of sequences in that set.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.