Where can I download the latest BED file for the Oncomine Breast cfDNA Assay?
The latest BED files can be downloaded from the Oncomine Breast cfDNA Assay product page at https://www.thermofisher.com/order/catalog/product/A31183.
I created a planned Oncomine Breast cfDNA Assay run in my Torrent Suite Software but cannot see it on my Ion GeneStudio S5 Series system. Why is this?
It is possible that there is a communication problem between the Ion GeneStudio S5 Series system and the server. We recommend shutting down the system, waiting for 2 minutes and then rebooting. If that does not work and if you are using the Ion S5 XL System, we recommend turning off the server and the system for 2 minutes. Then, attach a keyboard and monitor to the server and turn the server back on. Log in to the server and make sure you can connect to it via your web browser. Then, turn on the system. If the planned run does not appear, please contact Technical Support at techsupport@thermofisher.com for further assistance.
Why don't I see the Oncomine options in my Torrent Suite Software when planning an Oncomine Breast cfDNA Assay run?
You would need to use Torrent Suite Software version 5.2 or later in order to see Oncomine options.
Can I run the Oncomine Breast cfDNA Assay on the Ion S5 System or an Ion GeneStudio S5 Series system with a 510 or 520 chip?
The Oncomine Breast cfDNA Assay has only been validated on the Ion 530 and 540 chips at this time.
Can I run the Oncomine Breast cfDNA Assay on the Ion Proton System with a PI chip?
The Oncomine Breast cfDNA Assay has only been validated on the Ion 530 and 540 chips at this time.
Can I run the Oncomine Breast cfDNA Assay on the Ion PGM System using a 314, 316, or 318 chip?
The Oncomine Breast cfDNA Assay has only been validated on the Ion 530 and 540 chips at this time.
Can I use a spectrophotometer to quantify my Oncomine Breast cfDNA Assay library?
Library quantitation with a spectrophotometer may not be accurate as it will measure all DNA present instead of just the samples with both barcoded adapters attached.
The Oncomine Breast cfDNA Assay was left at 4 degrees C for more than an hour after target amplification. Can I still use it?
The PCR products can be stored at 4 degrees C for up to one hour.
Can I use single-cell DNA in the Oncomine Breast cfDNA Assay?
No. The recommended input amount is 20 ng of cfDNA but successful libraries have been made from 1-50 ng of cfDNA.
Why can't I use a spectrophotometer to quantitate my cfDNA samples for the Oncomine Breast cfDNA Assay?
We do not recommend using spectrophotometer methods for quantifying cfDNA because they are not reliable with low concentrations of nucleic acids. Use of these methods can result in overestimation of the concentration of sample, under-seeding of the target amplification reaction, and low library yields.
What is the best way to quantitate my cfDNA samples for use in the Oncomine Breast cfDNA Assay?
We recommend using the Qubit dsDNA HS Assay Kit (Cat. No. Q32851) for quantifying the cfDNA samples.
Which DNA isolation kit do you recommend for the Oncomine Breast cfDNA Assay?
We recommend using the MagMAX Cell-Free DNA Isolation Kit (Cat. No. A29319), which was included in-house during the Oncomine cfDNA workflow verification testing. Please follow the Alternate protocol for isolation of higher concentration cfDNA found in Appendix B of the MagMAX Cell-Free DNA Isolation User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0014327_A29319_magmax_cellfree...lasms_ug.pdf).
Where can I find the sequences of the primers used in the Oncomine Breast cfDNA Assay?
The primer sequences for the Oncomine Breast cfDNA Assay are proprietary.
What genes are covered in the Oncomine Breast cfDNA Research Assay v2?
The Oncomine Breast cfDNA Research Assay v2 covers the following genes:
- ALT1
- ERBB3
- KRAS
- CCND1
- ESR1
- PIK3CA
- EGFR
- FBXW7
- SF3B1
- ERBB2
- FGFR1
- TP53
More information on the coverage and the difference between versions 1 and 2 of the assay can be found in the flyer: Liquid Biopsy Cell Free Research Assays (https://assets.thermofisher.com/TFS-Assets/CSD/Flyers/liquid-biopsy-cell-free-research-assays-flyer.pdf).