Connector Cables for Automated Thermal Cycler (ATC)
Connector Cables for Automated Thermal Cycler (ATC)
Applied Biosystems™

Connector Cables for Automated Thermal Cycler (ATC)

Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
A314821-m connector cable (flexible)
A314833-m connector cable (flexible)
A3148510-cm connector cable (rigid)
Catalog number A31482
Price (USD)
122.00
Each
Add to cart
Format:
1-m connector cable (flexible)
Price (USD)
122.00
Each
Add to cart
Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.

For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you've come to expect from Applied Biosystems instruments. Small, easy-to-integrate, with a compatible format, our Automated Thermal Cycler enables hands-free PCR results.

In order to better align with lab design and demands, a longer or shorter cable may be utilized to optimize workflow.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)PCR
For Use With (Equipment)Automated Thermal Cycler (ATC)
Format1-m connector cable (flexible)
Max. Heating Rate (Thermal Block)2.8°C/sec. (Block), 1.6°C/sec. (Sample)
Quantity1 cable
Product TypeConnector Cable
Unit SizeEach