Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plates with Barcode, red
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plates with Barcode, red

The Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plate is optimally designed for use with the Applied Biosystems™ Automated ThermalRead more
Have Questions?
Catalog NumberQuantity
A3172950 plates
Catalog number A31729
Price (USD)
377.00
Each
Quantity:
50 plates
Price (USD)
377.00
Each
The Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plate is optimally designed for use with the Applied Biosystems™ Automated Thermal Cycler (ATC). These full-skirted reaction plates are manufactured using a durable polycarbonate frame, making them ideal for easy handling with robotic arms and other automated system features. The thin-wall construction of the plate allows for efficient heat transfer and optimal fit in thermal cycler blocks. The black alpha- numeric labeling provides easy-to-read well identification during sample preparation. The MicroAmp EnduraPlate Optical 96-Well Full-Skirted Plate is the only 96-well plate recommended for use with the Automated Thermal Cycler (ATC) in the automated PCR workflow process.

Key features include:
• Designed exclusively for use with Automated Thermal Cycler (ATC) (Cat No. A31486)
• Full-skirted polycarbonate frame for easy robotic handling and plate stacking/loading
• 3-way barcodes on alpha, opposite alpha, and opposite numeric sides to improve plate reading efficiency
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and automated work processes
• Certified free of DNA, RNase, and PCR inhibitors
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete automated workflow needs

Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Packaging50 Plates
Product TypeMicroplate
Quantity50 plates
Shipping ConditionRoom Temperature
Skirt StyleSkirted
Total Volume (Metric) Well0.2 mL
Volume (Metric) Working0.1 mL
BarcodeIncludes Barcode
ColorOptical, Red
MaterialPolycarbonate, Polypropylene
No. of Wells96
Product LineMicroAmp
Unit SizeEach
Contents & Storage
Store at room temperature.