Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, SYBR™ dyes), 96-well
Applied Biosystems™

Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, SYBR™ dyes), 96-well

The Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, SYBR™ dyes), 96-well is a specialized calibration tool to establish pure dye spectra and multicomponent values for four fluorescent dyes: FAM, VIC, ROX, and SYBR.
Have Questions?
Catalog NumberQuantity
A319071 Plate
Catalog number A31907
Price (USD)
-
Quantity:
1 Plate

The Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, SYBR™ dyes), 96-well is a specialized calibration tool to establish pure dye spectra and multicomponent values for four fluorescent dyes: FAM, VIC, ROX, and SYBR. This multi-dye plate offers significant operational efficiency by enabling simultaneous calibrations of multiple dyes within a single 96-well format, substantially reducing the effort and time traditionally required for individual dye calibration procedures.

Features:

  • Compatible with QuantStudio 5 Dx and QuantStudio 7 Pro Dx 96-well real-time PCR systems
  • Multiple Dye Compatibility: Supports simultaneous detection using multiple fluorescent dyes, enabling multiplex assay capabilities for enhanced analytical throughput

Simultaneous Multi-Dye Calibration Capability

This qPCR-based calibration system advances fluorescent dye calibration by enabling simultaneous calibration of multiple dyes in a single plate, reducing the time and effort required compared to traditional sequential methods.

Single-Plate Format Optimizes Resource Utilization

The single-plate format improves efficiency by enabling comprehensive multi-dye calibrations instead of multiple plates, while also reducing inter-plate variability to enhance calibration consistency.

Usage Guidelines and Technical Requirements

Proper use of this spectral calibration system requires following the procedures outlined in the instrument user guide to enable optimal performance, validated calibration, and compliance with quality standards. Users should consult the complete instrument documentation to achieve maximum performance and maintain compliance with established quality standards for qPCR spectral calibration procedures.

For In Vitro Diagnostic Use.
Specifications
Calibrated DyeFAM, VIC, ROX, SYBR Green
Calibration TypeSpectral Calibration Plate
For Use With (Application)Real-Time PCR (qPCR)
For Use With (Equipment)QuantStudio 5 Dx and QuantStudio 7 Pro Dx
Plate Compatibility96-well
Quantity1 Plate
Shipping ConditionDry Ice
Product TypeDye Spectral Calibration Plate
Unit SizeEach
Contents & Storage
Store at -20°C ±5°C.