Spectral Calibration Plate 2 (ABY™, JUN™, MUSTANG PURPLE™), 96-well
Applied Biosystems™

Spectral Calibration Plate 2 (ABY™, JUN™, MUSTANG PURPLE™), 96-well

The Spectral Calibration Plate 2 (ABY™, JUN™, MUSTANG PURPLE™), 96-well is a specialized calibration tool to establish pure dye spectra and multicomponent values for three fluorescent dyes: ABY, JUN, MUSTANG PURPLE.
Have Questions?
카탈로그 번호수량
A319081 Plate
카탈로그 번호 A31908
제품 가격(KRW)
450,000
온라인 행사
Ends: 31-Dec-2026
562,000
할인액 112,000
Each
수량:
1 Plate
제품 가격(KRW)
450,000
온라인 행사
Ends: 31-Dec-2026
562,000
할인액 112,000
Each

The Spectral Calibration Plate 2 (ABY™, JUN™, MUSTANG PURPLE™), 96-well is a specialized calibration tool to establish pure dye spectra and multicomponent values for three fluorescent dyes: ABY, JUN, MUSTANG PURPLE. This multi-dye plate offers significant operational efficiency by enabling simultaneous calibrations of multiple dyes within a single 96-well format, substantially reducing the effort and time traditionally required for individual dye calibration procedures.

Features:

  • Compatible with QuantStudio 5 Dx and QuantStudio 7 Pro Dx 96-well real-time PCR systems
  • Multiple Dye Compatibility: Supports simultaneous detection using multiple fluorescent dyes, enabling multiplex assay capabilities for enhanced analytical throughput

Simultaneous Multi-Dye Calibration Capability

This qPCR-based calibration system advances fluorescent dye calibration by enabling simultaneous calibration of multiple dyes in a single plate, reducing the time and effort required compared to traditional sequential methods.

Single-Plate Format Optimizes Resource Utilization

The single-plate format improves efficiency by enabling comprehensive multi-dye calibrations instead of multiple plates, while also reducing inter-plate variability to enhance calibration consistency.

Usage Guidelines and Technical Requirements

Proper use of this spectral calibration system requires following the procedures outlined in the instrument user guide to enable optimal performance, validated calibration, and compliance with quality standards. Users should consult the complete instrument documentation to achieve optimum performance and maintain compliance with established quality standards for qPCR spectral calibration procedures.

For In Vitro Diagnostic Use.
사양
보정된 염료ABY, JUN, MUSTANG PURPLE
보정 유형Spectral Calibration Plate
용도(애플리케이션)Real-Time PCR (qPCR)
용도(장비)QuantStudio 5 Dx and QuantStudio 7 Pro Dx
플레이트 호환성96-well
수량1 Plate
배송 조건Dry Ice
제품 유형Dye Spectral Calibration Plate
Unit SizeEach
구성 및 보관
Store at -20°C ±5°C.