Applied Biosystems™

TaqMan™ Vaginal Microbiota Extraction Control

Catalog number: A32039
Applied Biosystems™

TaqMan™ Vaginal Microbiota Extraction Control

Catalog number: A32039
Catalog Number
A32039
Unit Size
3 pellets
Price (USD)
948.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A320393 pellets
948.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
The TaqMan Vaginal Microbiota Extraction Control contains yeast-transformed cells in lyophilized pellets. These cells contain a linearized multi-target plasmid that includes amplicons for each of the qualified TaqMan Vaginal Microbiota assays, as well as control prokaryotic 16S rRNA and human RNase P RPPH1 gene sequences. The TaqMan Vaginal Microbiota Extraction Control can be used as a convenient training tool to learn and practice the protocol for isolating DNA from vaginal samples.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition
Ambient
Product Line
TaqMan™
Label or Dye
Unconjugated

Contents & Storage

3 pellets, 106 plasmid copies/pellet
Store (at 2°C to 8°C.)

Figures

Frequently asked questions (FAQs)

Citations & References