본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
인용 및 참조 문헌 (1)
Invitrogen™
LentiArray™ CRISPR Positive Control Lentivirus, human HPRT
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray™ CRISPR Positive Control자세히 알아보기
Have Questions?
보기 방식 변경
카탈로그 번호
수량
A32056
100 μL
A32829
1 mL
2 개 옵션
카탈로그 번호 A32056
제품 가격(KRW)
564,400
온라인 행사
Ends: 30-Sep-2026
664,000
할인액 99,600 (15%)
Each
수량:
100 μL
제품 가격(KRW)
564,400
온라인 행사
Ends: 30-Sep-2026
664,000
할인액 99,600 (15%)
Each
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice.
For Research Use Only. Not for use in diagnostic procedures.
사양
제어 유형Positive Control
배달 유형Lentiviral
용도(애플리케이션)CRISPR gene knockout
제품라인LentiArray
제품 유형CRISPR Positive Control Lentivirus
수량100 μL
배송 조건Dry Ice
프로모터U6
종Human
Transfection TechniqueLentiviral transduction
Unit SizeEach
구성 및 보관
1 x 100 μL. Store (at -68°C to -85°C.)
인용 및 참조 문헌 (1)
인용 및 참조 문헌
Abstract
A functional screening of the kinome identifies the Polo-like kinase 4 as a potential therapeutic target for malignant rhabdoid tumors, and possibly, other embryonal tumors of the brain.
Authors:Sredni ST, Suzuki M, Yang JP, Topczewski J, Bailey AW, Gokirmak T, Gross JN, de Andrade A, Kondo A, Piper DR, Tomita T
Journal:Pediatr Blood Cancer
PubMed ID:28398638
Malignant rhabdoid tumors (MRTs) are deadly embryonal tumors of the infancy. With poor survival and modest response to available therapies, more effective and less toxic treatments are needed. We hypothesized that a systematic screening of the kinome will reveal kinases that drive rhabdoid tumors and can be targeted by specific ... More