TrueMark™ OpenArray™ Vaginal Microbiota Training Plate
Applied Biosystems™
TrueMark™ OpenArray™ Vaginal Microbiota Training Plate
The TrueMark OpenArray Vaginal Microbiota Training Plate is a fixed-content panel containing 16 of the qualified TaqMan Vaginal Microbiota assays,Read more
Have Questions?
Catalog NumberQuantity
A320861 Plate
Catalog number A32086
Price (MXN)
-
Quantity:
1 Plate
The TrueMark OpenArray Vaginal Microbiota Training Plate is a fixed-content panel containing 16 of the qualified TaqMan Vaginal Microbiota assays, as well as assays for control prokaryotic 16S rRNA and human RNase P RPPH1 genes.
This pre-formatted inventoried plate is designed to offer a convenient and cost-effective alternative for users to learn the workflow prior to using custom OpenArray plates for vaginal and urogenital microbiota investigations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatOpenArray Plate
Product TypeVaginal Microbiota Training Plate
Quantity1 Plate
Shipping ConditionDry Ice
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at -5° to -30°C.