Alexa Fluor™ 488 Streptavidin, 5 nm colloidal gold conjugate
Invitrogen™

Alexa Fluor™ 488 Streptavidin, 5 nm colloidal gold conjugate

Alexa Fluor™ 488 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin hasRead more
Have Questions?
Catalog NumberQuantity
A32360
also known as A-32360
0.5 mL
Catalog number A32360
also known as A-32360
Price (USD)
436.00
Each
Quantity:
0.5 mL
Price (USD)
436.00
Each
Alexa Fluor™ 488 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor™ 488 Streptavidin Conjugates:
• Alexa Fluor™ 488 streptavidin conjugate has Ex/Em maxima of ∼ (495/519)
• Bright, photostable fluorescence
• High solubility in aqueous solutions
• Available in multiple colors
• Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

Properties of Alexa Fluor™ Dyes
Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Related Links:

Learn more about Avidin-Biotin Detection

Learn more about Alexa Fluor™ Dyes

Find out about other Labeled Streptavidin Conjugates

Read Avidin and Streptavidin Conjugates-Section 7.6 in the Molecular Probes Handbook
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration30 μg/mL
Label or DyeAlexa Fluor Dyes
Product TypeStreptavidin Conjugate (fluorescent)
Quantity0.5 mL
Shipping ConditionWet Ice
ConjugateAlexa Fluor 488, FluoroNanogold
FormLiquid
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?

One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.