What mounting media should I use with samples labeled with Alexa Fluor Plus secondary antibodies?
All Alexa Fluor Plus secondary antibodies are compatible with the SlowFade Diamond and ProLong Diamond Antifade Mountants.