Product Image
Applied Biosystems™
Extra MagMAX FFPE DNA/RNA Ultra Reagents
This package includes supplemental reagents for use with the MagMAX FFPE DNA/RNA Ultra Kit (Cat. No. A31881). These additional reagentsRead more
Have Questions?
Catalog NumberQuantity
A3279610 mL kit
Catalog number A32796
Price (USD)
279.65
Online exclusive
314.00
Save 34.35
Each
Quantity:
10 mL kit
Price (USD)
279.65
Online exclusive
314.00
Save 34.35
Each
This package includes supplemental reagents for use with the MagMAX FFPE DNA/RNA Ultra Kit (Cat. No. A31881). These additional reagents consist of protease digestion buffer, binding solution, and DNA wash buffer and are specifically for use when processing >40 um sections. See the User Guide for details.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeFFPE DNA/RNA Ultra Reagent
Quantity10 mL kit
Sample TypeFFPE, Fixed Samples
Shipping ConditionApproved for shipment at Room Temperature.
TargetGenomic DNA, Total RNA
Unit SizeEach
Contents & Storage
• 10 mL protease digestion buffer, store at room temperature
• 38.5 mL binding solution, store at room temperature
• 38.5 mL DNA wash buffer, store (at room temperature)
Frequently asked questions (FAQs)
Is there a limit to the size of curls that can be processed with the MagMAX FFPE DNA/RNA Ultra Kit (Cat. No. A31881)?
There is no specific upper limit to curl size for use with the MagMAX FFPE DNA/RNA Ultra kit (Cat. No. A31881). If all of the tissue is able to be submerged in the 200 µL of buffer, then the large volume of protocol should work. Please keep in mind when processing sections that are thicker than 40 µm, additional digestion buffer, binding solution, and wash buffer, may need to be purchased separately (Cat. No. A32796).