Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
Fisher Chemicals

Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™

H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
5 L
Packaging:
View all
Plastic Coated Glass bottle
Catalog number A3280PB17
also known as A/3280/PB17
HSN 28142000
Price (INR)
6,773.00
Each
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (INR)
6,773.00
Each
Specifications
pH12
ColorColorless
CAS1336-21-6
Molecular FormulaH5NO
MDL NumberMFCD00066650
View more
H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N