MagMAX™ CORE Lysis Solution
Applied Biosystems™
MagMAX™ CORE Lysis Solution
This MagMAX CORE Lysis Solution is made available as a supplement to that included in the MagMAX CORE Nucleic AcidWeitere Informationen
Have Questions?
KatalognummerMenge
A3283750 ml
Katalognummer A32837
Preis (EUR)
147,00
Each
Menge:
50 ml
Preis (EUR)
147,00
Each
This MagMAX CORE Lysis Solution is made available as a supplement to that included in the MagMAX CORE Nucleic Acid Purification Kit (Cat. No. A32700). This proprietary buffer solution is used to break open cells in preparation for downstream molecular detection applications such as real time PCR, RT-PCR, and sequencing applications.
For Research Use Only
Specifications
Zur Verwendung mit (Anwendung)Echtzeit-PCR, Sequenzierung
Hochdurchsatz-KompatibilitätGeeignet für hohe Durchsätze
ProduktlinieMagMAX
Menge50 ml
ProbentypBakterien, Blut, DNA, Liquid Samples (e.g. Serum), RNA, Gewebe, Virus
VersandbedingungRaumtemperatur
ZielGenomic DNA, Pathogen DNA/RNA, Total Nucleic Acids, Viral DNA/RNA
Unit SizeEach
Inhalt und Lagerung
1 x 50 mL MagMAX CORE Lysis Solution
Store at room temperature.