Pierce Protease Inhibitor Tablets - FAQs
Pierce Protease Inhibitor Tablets - FAQs
View additional product information for Pierce Protease Inhibitor Tablets - FAQs (A32963)
9 product FAQs found
Do Pierce Protease Inhibitor Tablets (Cat. No. A32963) dissolve quickly?
Pierce Protease Inhibitor Tablets (Cat. No. A32963) dissolve quickly to form a clear solution. We recommend dissolving 1 tablet in 50 mL of buffer or lysate. The protease inhibitor formulation is compatible with most detergent-based cell lysis reagents, including Pierce Cell Lysis solutions. Pierce Protease Inhibitor Tablets do not contain phosphatase inhibitors. If necessary, phosphatase inhibitors, such as Pierce Phosphatase Inhibitor Mini Tablets (Cat. No. A32957) can be added along with the protease inhibitor tablets.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?
We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What formats of protease inhibitors do you offer?
We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Do you offer any of the components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets individually?
Yes, the following components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are offered separately:
- AEBSF (Cat. No. 78431)
- Aprotinin (Cat. No. 78432)
- Bestatin (Cat. No. 8433)
- E-64 (Cat. No. 78434)
- Leupeptin (Cat. No. 78435)
- Pepstatin A (Cat. No. 78436)
- EDTA (Cat. No. 17892).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What are the concentrations of the various components of the protease and phosphatase inhibitor tablets and mini tablets?
The final concentration of each component in the protease and phosphatase inhibitor tablets and mini tablets is proprietary. We release only the identities of the inhibitors.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I store the reconstituted solution of protease, phosphatase and (protease and phosphatase) inhibitor tablet or mini tablet and if so, for how long?
The Pierce protease, phosphatase, as well as (protease and phosphatase) inhibitor tablets and mini tablets are stable for at least one year when stored at 4 degrees C, but we only recommend preparing fresh solutions just before needed. If you are interested in liquid protease or phosphatase inhibitors, please use our inhibitor cocktails (https://www.thermofisher.com/search/results?query=protease%20inhibitor%20cocktails&focusarea=Search%20All).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Which protein assay is compatible with the protease, phosphatase, as well as (protease and phosphatase) inhibitor tablets and mini tablets?
The Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are compatible with common protein assays such as BCA Protein Assay, 660 nm Protein Assay, Coomassie Plus (Bradford) Assay, and Detergent Compatible Bradford Assay.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
When do I need to use a protease or phophatase inhibitor tablet or mini tablet that contains EDTA?
Metalloproteases bind metal ions in their active sites. EDTA is a metalloprotease inhibitor that acts by chelating metal ions that are required by these proteases for activity. We recommend using the protease or phosphatase inhibitor tablet or mini tablet containing EDTA if any metalloprotease is likely to adversely affect the functioning of the target protein. However, it is important to avoid using EDTA when the downstream application involves metal chelation affinity purification or if the target protein requires metals ions for proper function.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Which protease and phosphatase inhibitor tablets do you offer?
The Pierce protease, phosphatase, as well as (protease and phosphatase) inhibitors are offered as mini tablets, with or without EDTA. Each mini tablet is sufficient for 10 mL of solution and is suitable for reagent preparation prior to protein extraction for cultured cells and tissues. In addition, the Pierce protease inhibitor tablets are offered with or without EDTA and each tablet is sufficient for 50 mL of solution.
Here are the catalog numbers:
-Pierce Protease Inhibitor Mini Tablets (Cat. No. A32953)
-Pierce Protease inhibitor Mini Tablets, EDTA-free (Cat. No. A32955)
-Pierce Phosphatase Inhibitor Mini Tablets (Cat. No. A32957)
-Pierce Protease and Phosphatase Inhibitor Mini Tablets (Cat. No. A32959)
-Pierce Protease and Phosphatase Inhibitor Mini Tablets, EDTA-free (Cat. No. A32961)
-Pierce Protease Inhibitor Tablets (Cat. No. A32963)
-Pierce Protease Inhibitor Tablets, EDTA-free (Cat. No. A32965)
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.