Thermo Scientific™ S1 Pipet Filler and Nunc™ Serological Pipettes
Thermo Scientific™

Thermo Scientific™ S1 Pipet Filler and Nunc™ Serological Pipettes

This combination of Thermo Scientific™ S1 Pipet Filler and Nunc™ Serological Pipettes offers a convenient start to pipetting productivity in cell culture and large volume pipetting applications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColorVolumen (métrico)
A33234Blanco10 mL
A33240Azul25 mL
A33237Transparente10 mL
A33242Transparente25 mL
A33236Verde10 mL
A33241Verde25 mL
A33238Rojo10 mL
A33243Rojo25 mL
Número de catálogo A33234
Precio (USD)
-
Color:
Blanco
Volumen (métrico):
10 mL

This combination of Thermo Scientific™ S1 Pipet Filler and Nunc™ Serological Pipettes offers a convenient start to pipetting productivity in cell culture and large volume pipetting applications.

The S1 Pipet Filler is a motorized filler with these features:

  • LCD display with speed and battery indicators
  • Separate speed control for aspiration and dispensing
  • Lithium-ion battery
  • Industry-leading 2-year warranty with web registration
  • A choice of five colors (white, blue, green, clear, red)

Nunc Serological Pipettes are disposable and individually wrapped pipettes with these features:

  • 100% virgin polystyrene for maximum clarity
  • Certified non-pyrogenic
  • Bold, clear graduations with easy-to-read scale markings
  • Non-dripping tip for pipette precision
  • Color-coded packaging for ease in sorting and selecting by size
  • Individually wrapped; paper/plastic or plastic film; peel or pop through for easy opening
  • Bulk packed for frequent use; reduces packaging waste
  • A choice of two sizes (10 mL and 25 mL)
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
ModeloS1
Condiciones de envíoTemperatura ambiente
ColorBlanco
Línea de productosNunc
Volumen (métrico)10 mL
Unit Size1 filler/200 pipettes
Contenido y almacenamiento
One S1 Pipet Filler and one case of 200 Nunc Pipettes