MicroSEQ™ ID Ultra Sequencing Clean-up Plates Kit - FAQs
MicroSEQ™ ID Ultra Sequencing Clean-up Plates Kit - FAQs
View additional product information for MicroSEQ™ ID Ultra Sequencing Clean-up Plates Kit - FAQs (A33245)
8 product FAQs found
How long is the MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No. A33245) stable for?
The product is guaranteed stable for one year when stored properly at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Can I store the MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No. A33245) at room temperature (20-24 degrees C)?
Yes. The kit contents are stable when stored in their original packaging at room temperature for up to 4 weeks. However, the recommended storage condition is at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Will the MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No. A33245) be compromised if I store it below –20 degrees C/frozen?
Unfortunately, frozen plates cannot be used. Continued use will result in total sample loss.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
How many sequencing samples is the MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No. A33245) sufficient for?
The MicroSEQ ID Ultra Sequencing Clean-up Plates Kit is sufficient for 192 sequencing samples.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What are the catalog numbers for the MicroSEQ ID Ultra Sequencing Clean-up Kits?
Here the kits with the catalog numbers:
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)
- MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No.A33245)
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is included in the MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No. A33245)?
The kit consists of two 96-well plates suitable for 192 sequencing samples.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is the minimum reaction volume for use of the MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No. A33245)?
The minimum reaction volume is 10 µL. The reaction volume should be brought up to at least 10 µL with distilled water before adding to the 96-well plate.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is the storage condition for the MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No. A33245)?
The kit is stable when stored for 12 months at 4 degrees C or for 4 weeks at ambient temperature (20-24 degrees C).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.