Can Phasemaker Tubes be used for isolation of DNA or protein?
No. Phasemaker tubes are only validated for RNA extraction.
Search
View additional product information for TRIzol™ Plus RNA Purification Kit and Phasemaker™ Tubes Complete System - FAQs (A33254)
1 product FAQs found